Student Plasticware Set
# de catalogue 470020-964
Fournisseur: United Scientific Supplies
Spécifications
- Description:Student Plasticware Set
- Cat. no.:470020-964
- Supplier No.:PLKIT20
Spécifications
A propos de cet article
Excellent started set with a wide variety of plasticware.
- Comes in a convenient storage case
- Safe plastic so no risk of injuries from broken glass
This affordable assortment of labware contains some of the most commonly used pieces. The plastic set comes with 20 items packed in a plastic storage box. The set includes:2 Beakers, 100 mL and 250 mL2 Graduated cylinders, 10 mL and 100 mL1 Funnel, 65 mm top dia.1 Drop dispenser bottle3 Test tubes4 Reagent bottles1 Wash bottle6 Pipets
Customers Who Bought This Also Bought