Student Pack Glass Slides and Cover Glasses
# de catalogue 470152-826
Fournisseur: Avantor
Spécifications
- Description:Student pack, glass slides and supplies
- Cat. no.:470152-826
- Supplier No.:WARD470152-826
Spécifications
A propos de cet article
Begin making your own slides!
- Explore slidemaking with this starter pack
- Hands-on learning
This pack includes 12 standard slides of 1×3" (25×75 mm), 12 No. 2 thickness of 0.2 mm (22 mm square) cover glasses, 12 slide label stickers, and a slide storage box.
Customers Who Bought This Also Bought