ContactUsLinkComponent
Student Molecular Model Set
Student Molecular Model Set
# de catalogue 470123-040
STUDENT MOLECULAR MODEL SET
Student Molecular Model Set
# de catalogue 470123-040
Spécifications
  • Description:
    Student Molecular Model Set
  • Cat. no.:
    470123-040
  • Supplier No.:
    FOM-303
Spécifications
A propos de cet article
Kit uses painted styrofoam balls for atoms and toothpicks and pipe cleaners for bonds. Students can learn concepts of the fundamentals of chemical bonding, reactivity and isomerism from this kit.
Kit contains four black, three red, four green, two blue and 15 white balls, toothpicks, pipe cleaners and a sp3 bonding jig.
Customers Who Bought This Also Bought