Student Double-Wall Calorimeter
# de catalogue 470110-282
Fournisseur: VWR International
Spécifications
- Description:Student Double-Wall Calorimeter
- Cat. no.:470110-282
- Supplier No.:DWCAL
Spécifications
A propos de cet article
The Student Double-Wall Calorimeter is an inexpensive necessity for your experiments.
- Aluminum Construction
- Double-Wall
- Styrofoam Can Width: 1.7 cm
- Reservoir Holds Maximum of 150 ml
- Clear Plastic Lid Has Three Holes
The aluminum, double-wall calorimeter is insulated with a 1.7 cm thick Styrofoam can and a plastic insulator ring.
The reservoir holds a maximum of 150 ml, and the clear plastic lid has three access holes.
Ordering information: This calorimeter includes cork with a hole for holding a thermometer, stirrer and a lid for the third access hole.
Customers Who Bought This Also Bought