Ward's® Student Clinometer
# de catalogue 470018-762
Fournisseur: Avantor
Spécifications
- Description:WARD’S Student Clinometer
- Cat. no.:470018-762
- Supplier No.:WARD470018-762
Spécifications
A propos de cet article
Economical enough to distribute to all students.
- Aligned with GLOBE protocols
The protractor quickly measures angles of dipping rock units or fracture planes — just take a reading where the weighted thread intersects the protractor. It features black printing on a white plastic face in 1° increments up to 90° on each side of zero. It comes with a case. Size: 4 3/8"L x 2 1/2"W.
Customers Who Bought This Also Bought