Stromatolite (Microbialite)
# de catalogue 470154-726
Fournisseur: ADAM MOORE MO
Spécifications
- Length:1–2"
- Description:Stromatolite (Microbialite)
- Cat. no.:470154-726
- Supplier No.:3533105
Spécifications
A propos de cet article
Unusual well-formed sedimentary bodies believed to have formed from the aggregates of algal organisms and cyanobacteria in ancient waters.
- Cyanobacteria
Interesting concentric growth structures. Limited Edition. Morocco.
Customers Who Bought This Also Bought