Streptococcus pyogenes Slide
# de catalogue 470177-414
Fournisseur: Avantor
Spécifications
- Description:Streptococcus pyogenes Slide
- Cat. no.:470177-414
- Stain:Gram (+)
- Supplier No.:WARD470177-414
- Orientation:Smear
Spécifications
A propos de cet article
Part of the human skin flora
- Identifying features clearly distinguishable
- Rigorous quality control standards
Spherical cells in long chains. Found in the human respiratory tract or in human blood.
Customers Who Bought This Also Bought