Stream Table Recirculating Pump
# de catalogue 470007-780
Fournisseur: Avantor
Spécifications
- Description:Stream Table Recirculating Pump
- Cat. no.:470007-780
- Supplier No.:WARD470007-780
Spécifications
A propos de cet article
Regulated, Constant Supply of Water
Much easier and less messy than adding water by hand, the 1/70 hp pump circulates up to 300 gallons per hour. It comes with a reservoir and mounting bracket. 115 VAC, 60 Hz. Reservoir size: 14"L x 12"W x 4"H.
Customers Who Bought This Also Bought