Ward's® Stream Table Cart
# de catalogue 470020-366
Fournisseur: PETTINGER SHEET & METAL
Spécifications
- Description:WARD’S Stream Table Cart
- Cat. no.:470020-366
- Supplier No.:158565
Spécifications
A propos de cet article
Top-Quality Steel Construction
Specifically designed to hold WARD’S Standard Stream Table or Deluxe Stream Table. The large, locking casters enhance mobility, and the lower shelf provides convenient storage for accessories. It comes preassembled. Size: 64"L x 24"W x 36"H. Weight: 75 lbs.
Customers Who Bought This Also Bought