Straight Connector
# de catalogue 470174-236
Fournisseur: Dynalon
Spécifications
- Description:Straight Connector
- Cat. no.:470174-236
- Supplier No.:227245
Spécifications
A propos de cet article
They are polypropylene with a ribbed design for a tight fit. Each fits tubing 1/4 – 5/6" Package of ten.
Customers Who Bought This Also Bought