Straight Connecting Tubes
# de catalogue 470174-240
Fournisseur: Dynalon
Spécifications
- Description:Tube, Connecting, Straight, 4-6 mm OD
- Cat. no.:470174-240
- Supplier No.:227235
Spécifications
A propos de cet article
These autoclavable straight connectors are molded of polypropylene and have serrations to hold tubing tightly and prevent slipping. They have excellent chemical and temperature resistance.
Customers Who Bought This Also Bought