Stoddard Test Tube Clamps with Finger Grips
# de catalogue 470176-124
Fournisseur: PRECISION MOULDED POLYMERS LIMITED
Spécifications
- Description:Stoddard Test Tube Clamps with Finger Grips
- Cat. no.:470176-124
- Supplier No.:M73602
Spécifications
A propos de cet article
Ideal for student use, these clamps hold test tubes measuring up to 25 mm firmly and can be manipulated easily. Made from brass.
One-piece construction with an overall length of 13 cm (5 inches).With molded finger grips to help students work with test tubes.
Package of six.
Customers Who Bought This Also Bought