Steel Horseshoe Magnet
Catalog # 470149-496
Supplier: VWR International
Specifications
- Description:Steel Horseshoe Magnet
- Cat. no.:470149-496
- Supplier No.:P40040/3
Specifications
About this item
Classically styled chrome steel magnets are large, easy to work with and affordably priced.
- Painted bright red with silver poles. Keepers included
- Length: 6.3 cm
- Chrome: 3.5%
Customers Who Bought This Also Bought