Steel Crucibles
Supplier: United Scientific Supplies
Certificates
About this item
Excellent Corrosion Resistance.
- Constructed of 1-piece steel sheet
- Uniform wall thickness
Unibody construction from Type 304 stainless steel. Can withstand temperatures from –180 to +800 °C. Covers included.
Customers Who Bought This Also Bought