Steel Bar Magnet
# de catalogue 470149-502
Fournisseur: VWR International
Spécifications
- Description:Steel Bar Magnets, 20 cm long
- Cat. no.:470149-502
- Supplier No.:P40022/6
Spécifications
A propos de cet article
Standard Magnets for Physics Labs
- 19mm x 6mm cross-section
Steel is the simplest material for permanent magnets as well as the most economical.
Customers Who Bought This Also Bought