ContactUsLinkComponent
Steam Engine Model
Steam Engine Model
# de catalogue 470120-694
Fournisseur:  VWR International
MODEL OF STEAM ENGINE
Steam Engine Model
# de catalogue 470120-694
Fournisseur:  VWR International
Spécifications
  • Description:
    Steam Engine Model
  • Size:
    38 x 12 x 16 cm
  • Cat. no.:
    470120-694
  • Supplier No.:
    PH0483
Spécifications
A propos de cet article
Demonstrate Principles of the First Commercially Produced Steam Engine.
No longer in frequent use, the steam engine was one of the first inventions which sparked an industrial revolution. This all metallic model enables you to show how this once revolutionary engine works well still today. The metal cylinder is section cut and covered with a glass plate for easy viewing within. Working parts include a piston and slide valve, so link motion and reversing method can be observed. The movement of the wheel can even be demonstrated by blowing on it.
Customers Who Bought This Also Bought