Staurolite (Twin)
# de catalogue 470154-882
Fournisseur: ADAM MOORE MO
Spécifications
- Description:Staurolite (Twin) Display Mineral
- Cat. no.:470154-882
- Supplier No.:493804
Spécifications
A propos de cet article
Classic staurolite crystal twins in schistose matrix with well-defined crystal form. Average size: 2" x 3". Russia.
Customers Who Bought This Also Bought