Prepared Starch Agar Media
# de catalogue 470180-696
Fournisseur: IPM SCIENTIFIC, INC.
Numéro CAS:
Spécifications
- Packaging:Plates
- Description:Starch Agar
- Cat. no.:470180-696
- Supplier No.:880927
Spécifications
A propos de cet article
This media can be used to determine if a microorganism is capable of hydrolyzing starch as a source of energy for growth.
- Sterile, pre-prepared media; just inoculate and incubate.
- When used with an iodine solution, works as a test for alpha-amylase.
After the organism is plated onto the starch agar plate, an organism tests positive for alpha-amylase when, in the presence of an iodine solution, a blue-black color forms in the medium.
Each bottle pours approximately 10 Petri dishes. Simply melt in a microwave oven, water bath or autoclave. Once cool enough to handle, pour into sterile Petri dish plates
Customers Who Bought This Also Bought