Variable Standing Wave Generator
# de catalogue 470039-704
Fournisseur: INDUSTRIAL SUPPORT INC.
Small Business Enterprise
Spécifications
- Description:Variable Standing Wave Generator
- Cat. no.:470039-704
- Supplier No.:180185
Spécifications
A propos de cet article
An exciting, interactive exploration, this generator includes various mass attachments to investigate degrees of string tension in standing waves.
- Ø×L: 3×15 cm
- String length: 5.5 m
- Weight: 0.5 kg
The device features a battery-operated motor with an off balanced bolt attached to the shaft. When running, this motor creates a vibration in the string that sets up a standing wave along the entire length of string. The nuts on the bottom alow you to change the center of mass, and therefore change the frequency of the standing waves. The top eyebolt allows slotted masses to be attached, so the affect of string tension on the wavelength can also be investigated. Developed by Bruce Yeany.
Customers Who Bought This Also Bought