Stainless Steel Spoon/Spatula
Supplier: VWR International
Certificates
About this item
Constructed entirely of stainless steel.
One end is spoon shaped, 13 mm wide, and the other is flat, with square corners (8 mm wide).
Customers Who Bought This Also Bought