Stainless Steel Scalpel
# de catalogue 470005-454
Fournisseur: Dunrite Instruments
Spécifications
- Length:152 mm (6")
- Description:Stainless Steel Scalpel
- Material:Stainless Steel
- Cat. no.:470005-454
- Supplier No.:140875
Spécifications
A propos de cet article
The extra-heavy blade molded onto a one-piece stainless steel scalpel won’t break off, making it easier to cut muscle and cartilage. Blade: 1¹/₂".
Caution: These instruments are intended for training and educational use only. They are not intended for human or animal procedures.
Customers Who Bought This Also Bought