Stainless Steel Laboratory Carts
Fournisseur: Lakeside
Certificats
A propos de cet article
Valuable time-savers in the laboratory for moving and dispensing chemicals, apparatus, and supplies, the carts are made from corrosion resistant stainless steel for permanent beauty and easy cleaning.
Each has three shelves that measure 39 x 61 cm and each a 2.5 cm molding on three sides. There is a 30.5 cm clearance between each shelf.
Units are shipped assembled with 10cm ball bearing, swivel casters and handle. Size of each: 43 x 71 x 81 cm.
Customers Who Bought This Also Bought