Stainless Steel Guard Rail Utility Cart
# de catalogue 470128-734
Fournisseur: Lakeside
Spécifications
- Description:Stainless Steel Guard Rail Utility Cart
- Cat. no.:470128-734
- Supplier No.:526
Spécifications
A propos de cet article
Durable and Sanitary Construction
Transport fragile items without the worry of spills or breakage using this cart with protective guard rail. Integrated push handle allows for easy maneuverability in your lab, and an all-welded, durable stainless steel construction enables easy sanitation. Standard handle and leg bumpers are included to protect cart, walls and doorways from damage.
Customers Who Bought This Also Bought