Squirrel Skull
# de catalogue 470027-990
Fournisseur: SKULLS UNLIMITED
Spécifications
- Description:Squirrel Skull
- Height:1.25"
- Cat. no.:470027-990
- Length:2 .5"
- Width:1.25"
- Supplier No.:SM 450A
- No. of Parts:1
Spécifications
A propos de cet article
Customers Who Bought This Also Bought