Surgeon's Color Atlas Of Large Animal Anatomy
# de catalogue 470023-138
Fournisseur: BLACKWELL PUBLISHING
Spécifications
- Description:Surgeon's Color Atlas Of Large Animal Anatomy
- Cat. no.:470023-138
- No. of Pages:159
- Length:23 cm
- Width:30 cm
- Supplier No.:0683306731
Spécifications
A propos de cet article
Thomas Spurgeon et al
Original artwork and extraordinary accuracy combine with unique organization in this essential resource that highlights the structure of various animal species. Explore the anatomy of all the organ systems in numerous animals, including horse, sheep, goat, llama, alpaca, swine, and chicken. Both male and female of a given species are depicted on facing pages so that topographic anatomy is easy to compare.
Customers Who Bought This Also Bought