Specific Gravity Specimens Set
# de catalogue 470005-426
Fournisseur: ELECTRO MECHANICAL INSTRUMENTS
Spécifications
- Description:Specific Gravity Specimens Set
- Cat. no.:470005-426
- Supplier No.:1012
Spécifications
A propos de cet article
Includes a variety of materials for density experiments
- The set includes ten different substances, each with a approximate volume of 1 cubic centimeter
- Cubes are made of aluminum, copper, plastic, rubber, steel, brass and wood
- The set also includes a carbon cylinder, a lucite block and a cork block
Students should measure to verify the exact volume. The objects in the set consist of different shapes, materials and masses. Average densities for the various materials in this set are aluminum 3.11, copper 10.16, cork 0.23, lucite block 1.46, plastic 1.78, rubber 1.92, steel 9.10, wood 0.84, carbon cylinder 0.40 and brass 9.92 (all in g/cubic cm).
Customers Who Bought This Also Bought