Specific Gravity Blocks Set
# de catalogue 470006-214
Fournisseur: Avantor
Spécifications
- Description:Specific Gravity Blocks Set, Set of 3
- Cat. no.:470006-214
- Supplier No.:WARD470006-214
Spécifications
A propos de cet article
Teach your students properties of materials!
Add a balance and this set provides an effective package for small demonstrations or student work on specific gravity. The set consists of three 3.2 cm cubes (aluminum, brass, and iron) with hooks and our Catch Bucket and Overflow Can, plus complete instructions for use. Blocks are also available individually.
- The set consists of three 3.2 cm cubes (aluminum, brass, and iron) with hooks and our Catch Bucket and Overflow Can
- Includes complete instructions for use
Add a balance and this set provides an effective package for small demonstrations or student work on specific gravity. Blocks are also available individually.
Customers Who Bought This Also Bought