Ward's® Prepared Sordaria Mating Agar
Fournisseur: Culture Media & Supplies
Certificats
A propos de cet article
For the Study of Genetic Crossover
- Sterile- no autoclaving
- Convenient - prepared for you
- Prepared consistenly every time
Supports and enhances mating of Scordaria strains for genetic studies.
Ordering information: Some of these products contain living or perishable materials
Delivery: Prepared plates will ship via overnight or 2nd day delivery.
Prepared bottles and tubes do not require as much temperature regulation in-transit so we ship them via ground to save you the cost of freight. During checkout, please specify a delivery date for your specimens to arrive.
Prepared bottles and tubes do not require as much temperature regulation in-transit so we ship them via ground to save you the cost of freight. During checkout, please specify a delivery date for your specimens to arrive.
Customers Who Bought This Also Bought