Ward's® Sordaria Crossing-Over Flashcards
# de catalogue 470007-268
Fournisseur: PROFORMA ASSOCIATES
Spécifications
- Description:Sordaria Flash Cards, 25 Cards
- Cat. no.:470007-268
- Length:12.7 cm
- Width:7.6 cm
- Supplier No.:323377
Spécifications
A propos de cet article
Striking Images Capture Meiotic Recombination
- Perfect companion to any crossing over lab activity
Determine the percent of crossing over that occurs in Sordaria asci by using the different photomicrograph shown on each card. The set is a perfect companion to any crossing over lab activity or when you are unable to prepare your own slides.
Customers Who Bought This Also Bought