Sordaria Agar
# de catalogue 470175-878
Fournisseur: MICROLOGY INC
Spécifications
- Description:Prepared Sordaria Agar Bottles
- Cat. no.:470175-878
- Supplier No.:WARD470175-878
Spécifications
A propos de cet article
For growth and sporulation of ascomycetes, specifically Sordaria species.
Customers Who Bought This Also Bought