ContactUsLinkComponent
Sordaria Agar
Sordaria Agar
# de catalogue 470175-878
Fournisseur:  MICROLOGY INC
Sordaria Agar
# de catalogue 470175-878
Fournisseur:  MICROLOGY INC
Spécifications
  • Description:
    Prepared Sordaria Agar Bottles
  • Cat. no.:
    470175-878
  • Supplier No.:
    WARD470175-878
Spécifications
A propos de cet article
For growth and sporulation of ascomycetes, specifically Sordaria species.
Customers Who Bought This Also Bought