Somso® Woody Dicot Stem Section Model
# de catalogue 470029-852
Fournisseur: MARCUS SOMMER
Spécifications
- Description:Woody Dicot Stem Section
- Height:18 cm
- No. of Keyed Items:18
- Cat. no.:470029-852
- Length:30 cm
- Width:65 cm
- Supplier No.:BOS 21/1
- No. of Parts:1
Spécifications
A propos de cet article
Detailed Representation Of A Two-Year Tilia Twig
- Woody Growth Shown In 3 Traditional Sections
- 5 Year Manufacturers Warranty
Showing all the elements of wood structure in great detail, the model of a basswood twig is presented in transverse, radial, longitudinal, and tangential sections. The 350X enlargement depicts the section from the center of the twig to the epidermis and is ideal for comparison to the structure of the Herbaceous Monocot Stem Section Model.
Customers Who Bought This Also Bought