Somso® Paramecium Model
# de catalogue 470029-782
Fournisseur: MARCUS SOMMER
Spécifications
- Description:Model Paramecium, 33 keyed itms, 3 parts
- L×W:560×280 mm
- Cat. no.:470029-782
- Slide Type:Protists
- Supplier No.:ZOS 107
Spécifications
A propos de cet article
Two Accurate, Highly Detailed Models In One
- 5 Year Manufacturers Warranty
For an overall view, have your students examine the complete model of a whole Paramecium, enlarged 1,600X and mounted on a stand for easy viewing by the entire class. Sectioned longitudinally, the outer half can be detached to reveal macro- and micronuclei, a contractile vacuole, membranelles, myonemes, neuroneme network, food vacuoles, and formation of endo- and ectoplasm. The second model, the pellicle enlarged 6,500X, provides an up-close view of trichocysts and ciliary bending action illustrating a metachronal wave. Both models are mounted on a common base; the pellicle is mounted directly on the base, while the Paramecium is mounted on a stand.
Customers Who Bought This Also Bought