Somso® Chick Embryo Development Model Set
# de catalogue 470029-804
Fournisseur: MARCUS SOMMER
Spécifications
- Description:Chick Embryo Development Set
- No. of Keyed Items:168
- Cat. no.:470029-804
- Supplier No.:ZOS103ZOS103/5
- No. of Parts:6
Spécifications
A propos de cet article
Six-Model Set Illustrates Embryological Growth
- Chick Embryology From Fertilization To 96 Hours
- Individual Models Are Great For Student Review, Lab Stations And Practicals
- 5 Year Manufacturers Warranty
Present your students with a colorful, highly detailed, and greatly enlarged reproduction of the growth and development inside a chicken’s egg. Starting with the fertilized egg itself and continuing until the chick embryo begins to take shape, the six individual models work together to provide a complete picture of the first days of cell development. Sizes range from 16 x 16 x 45 cm to 35 x 23 x 39 cm.
Customers Who Bought This Also Bought