Solvent Spill Clean Up Kit
# de catalogue 470127-672
Fournisseur: Ward's Science
Spécifications
- Description:Solvent Spill Clean Up Kit
- Cat. no.:470127-672
- Supplier No.:IS5001
Spécifications
A propos de cet article
For Safe Disposal of Chemical Spills.
- Handy compact design
- Contains everything to safely contain the spill
Keep this kit on hand in your chemical storeroom or laboratory to be prepared for safe clean-up of inevitable solvent spills. Includes one bag of diatomaceous earth, two vinyl exam gloves, two 9" x 16" polybags, two blank shipping tags, twist ties, and a dust pan and brush set. A detailed instruction sheet/SDS is provided within the kit.
Customers Who Bought This Also Bought