Solderless Breadboard
# de catalogue 470101-274
Fournisseur: JA ASSOCIATES
Spécifications
- Description:Solderless Breadboard
- Cat. no.:470101-274
- Supplier No.:JAM JE21
Spécifications
A propos de cet article
This solderless breadboard is ideal for high-frequency and low-noise circuits.
Perform your electronic experiments on this tiny breadboard (8×5.5×1 cm) . Interconnect any components with 20-29 AWG (3 to 8 mm) wire.
Its highly durable nickel-plated contacts provide a long life and low resistance connections. An added feature is the adhesive backing, allowing you to secure it anywhere.
This breadboard has a specially designed spring clip that’s made to withstand over five thousand insertion cycles.
Customers Who Bought This Also Bought