Soil Thermometer
# de catalogue 470106-366
Fournisseur: Taylor Precision Products
Spécifications
- Description:Thermometer, Soil, Stainless Steel
- Cat. no.:470106-366
- Divisions:1 °C
- Range:0...80 °C
- Supplier No.:5976N
Spécifications
A propos de cet article
Determine Soil Temperature Quickly.
- Large visible display of optimal growing conditions
- Durable
- Aligned with GLOBE protocols
Use this stainless steel thermometer to test soil temperatures for planting. The dial, which is 6 cm in diameter, records temperatures from 0 to 80 °C and 20 to 180 °F. The 12.5 cm stem is long enough for most planting depths.
Customers Who Bought This Also Bought