ContactUsLinkComponent
Soil Reaction pH Tester
Soil Reaction pH Tester
# de catalogue 470021-094
Fournisseur:  Lovibond
Small Business Enterprise
SOIL REACTION PH TESTER KIT-200 TESTS
Soil Reaction pH Tester
# de catalogue 470021-094
Fournisseur:  Lovibond
Small Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Soil Reaction pH Tester
  • Cat. no.:
    470021-094
  • Supplier No.:
    694 (QB)
  • No. of Tests:
    200
Spécifications
A propos de cet article
Works Directly on a Soil Sample, Eliminating the Need for a Soil Extract
  • Colorimetric pH determination
  • Useable on any soil type
  • Includes instructions
Eliminate difficulties with soil extracts that may be turbid or colored by the soil itself. The special indicator, which works on any type of soil, exhibits easy-to-distinguish colors in a colorimetric reaction for comparison to a color chart to determine the soil’s pH for a range of pH 4–8.5. The boxed set includes all materials for 200 tests.
Customers Who Bought This Also Bought