Small Magnetic Compasses
# de catalogue 470005-402
Fournisseur: PRECISION MOULDED POLYMERS LIMITED
Spécifications
- Description:Small Magnetic Compasses
- Cat. no.:470005-402
- Supplier No.:WARD470005-402
Spécifications
A propos de cet article
Very inexpensive magnetic compasses are specially suited for tracing the magnetic fields of bar magnets. Each compass has a polished aluminum case and a beveled glass top.
- Ø: 20 mm
Packaging: Pack of 12.
Customers Who Bought This Also Bought