Slotted Microscope Slide Staining Dish
Catalog # 470020-694
Supplier: C & A Scientific
Specifications
- Width:2"
- Capacity:20
- Description:Slotted Microscope Slide Staining Dish
- Length:3"
- Cat. no.:470020-694
- Supplier No.:SD-10
Specifications
About this item
Durable Staining Dish with Less Mess
- Quality slide making supplies
- Slide staining tools
This deep covered glass dish lessens the risk of overflow. It will hold 20 slides back to back or ten single slides.
Customers Who Bought This Also Bought