Skim Milk Agar Plates
# de catalogue 470175-918
Fournisseur: Culture Media & Supplies
Numéro CAS:
Spécifications
- Description:Prepared Skim Milk Agar Plates
- Cat. no.:470175-918
- Supplier No.:WARD470175-918
Spécifications
A propos de cet article
For differentiation of organisms based on proteolysis of casein and coagulation.
Customers Who Bought This Also Bought