The Skeletal System Microslide
Fournisseur: SUPERTEK SCIENTIFIC LLC
Certificats
A propos de cet article
Investigate the structural system of the body
- Arrows help students locate important features
- Mounted in a clear plastic holder that protects it on both sides
Microslides consist of a strip of film with photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own microslides. A detailed text folder is included. Viewers are available separately.
Ordering information: Set includes Bone (cs, st, 150X); Membranous bone (cs, st, 120X); Bone narrow, left side normal (st, 400X); right side leukemia, (st, 500X); Bone fracture, arm (x-ray, 1/8X); Cartilage (st, 69X); Arthritis, hands (top, 1/6X); Arthritis, feet (x-ray, bottom, 1/6X); Ligament at a joint, left side (wm, 1/2X); right side, fibers, teased (200X); and Muscle-tendon-bone connections, left side (cs, 400X); right side (cs, 10X).
Customers Who Bought This Also Bought