ContactUsLinkComponent
Siren Disk
Siren Disk
# de catalogue 470005-720
Fournisseur:  Science First
ROTATOR UNIVERSAL
Siren Disk
# de catalogue 470005-720
Fournisseur:  Science First
Spécifications
  • Description:
    Universal Rotator
  • Cat. no.:
    470005-720
  • Supplier No.:
    ER-6A
Spécifications
A propos de cet article
Explore the relationship between acoustical tones and the physical circumstances that give rise to them
  • Perforated rotating disk
  • Made under computer control
  • 18 x 13 x 4.5 cm
  • 23 cm H
Can be used to investigate the resonance of tubes and containers and to explore the evolution of musical scales. Comes with a sounding tube and mandrel. We recommend mounting it on a standard lab rotator such as our Universal Rotator (shown here). Uniformly spaced holes are 8, 10, 12, 16, 24, 27, 30, 32, 36, 40, 45 and 48 holes per revolution.
Customers Who Bought This Also Bought