Simple Machines Set
# de catalogue 470017-812
Fournisseur: Learning Resources
Spécifications
- Description:Simple Machines Set
- Cat. no.:470017-812
- Supplier No.:LER 2442
Spécifications
A propos de cet article
Build 5 simple machines simultaneously!
- Simple machines
- Explore concepts of movement
- Easy assembly
Students can build a lever, pulley, inclined plane, wheel and axle, and wedge all at the same time with this set. These simple machines will enable students to explore effort, force, load, motion and distance as students modify machines with four included weights (two 5g and two 10 g) and eight included rubber bands. Students will develop a deeper understanding of how these five simple machines make work easier. There are labeled pieces which assemble easily and can be separated for storage. Set of 63 pieces includes Activity Guide. Grades 4+.
Caution: WARNING: CHOKING HAZARD - Small parts. Not for children under 3 years.
Customers Who Bought This Also Bought