Simple Form Tangent Galvanometer
# de catalogue 470006-690
Fournisseur: VWR International
Spécifications
- Description:Galvanometer, Simple Form Tangent
- Dimensions:16.5 x 15.2 x 19 cm
- Cat. no.:470006-690
- Supplier No.:PH0810
Spécifications
A propos de cet article
Simple and efficient.
- Investigate the magnetic field of an electric current
The simplicity of this design makes a very efficient unit for investigating the magnetic field of an electric current. Smaller than previous models, it supports a wire coil approximately 18 cm on a side with a horizontal masonite shelf intersecting its middle. Included are 335 cm of stranded copper wire and a large 4 cm diameter compass.
Customers Who Bought This Also Bought