Simple Columnar Epithelium, Mammal Kidney Slide
# de catalogue 470182-604
Fournisseur: Avantor
Spécifications
- Description:Simple Columnar Epithelium, Mammal Kidney Slide
- Stain:Hematoxylin and Eosin
- Supplier No.:WARD470182-604
- Orientation:Section
Spécifications
A propos de cet article
Illustrates tissue that lines most human organs
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal kidney. Shows secreting tubules.
Customers Who Bought This Also Bought