Simple Ciliated Columnar Epithelium
# de catalogue 470177-566
Fournisseur: Avantor
Spécifications
- Description:Simple Ciliated Columnar Epithelium
- Cat. no.:470177-566
- Stain:Hematoxylin and Eosin
- Supplier No.:WARD470177-566
- Orientation:Section
Spécifications
A propos de cet article
Cilia move mucus and other substances mainly within the respiratory and reproductive tracts
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Simple ciliated columnar epithelium demonstrated in the oviduct.
Customers Who Bought This Also Bought