Sulfide Indole Motility (SIM) Agar Tubes
Catalog # 470180-658
Supplier: Avantor
CAS Number:
Specifications
- Description:Prepared SIM Medium Tubes
- Cat. no.:470180-658
- Supplier No.:WARD470180-658
Specifications
About this item
For the differentiation of members of the Enterobacteriacaea based on H2S production, indole production, and motility.
Customers Who Bought This Also Bought