Side Desk for Instructors Lab Tables
Fournisseur: Diversified Woodcrafts
Certificats
A propos de cet article
This side desk is a great addition to the 5' and 8' Instructor's Desk for additional workspace and storage space.
- 3/4" Phenolic resin top
- Constructed of solid oak and oak veneers
- An earth-friendly chemical resistant UV finish
- Drawers are of dovetail construction and come with locks
- Features two shelves in the back of the pedestal
The side desk features one file drawer, three small drawers including two storage drawers, one pencil drawer and a pull-out writing board. The classroom side of the unit features a bookshelf with one fixed shelf. The unit can be attached to either the right or left side of the instructor desk.
File drawer W×D×H: 13.5×18×10.25"
Pencil drawer W×D×H: 19.5×18×3"
Storage drawer W×D×H: 13.5"×18"×4"
Writing board W×D×H: 14.5×20"
Shelf W×D×H: 14.5×7.5×11.25"; 40 lbs. per shelf
Certifications: SEFA compliant and MAS Certified Green.
Spécifications
- Catalog No:
- 470022-414
- Environmentally Preferable:
- Safer Human and Environmental Health
- Environmental certification:
- MAS Certified Green