Shower Flush System
# de catalogue 470039-360
Fournisseur: JOEL C. PALMER
Spécifications
- Description:Shower Flush System
- Cat. no.:470039-360
- Supplier No.:175710
Spécifications
A propos de cet article
Perform the monthly shower flushing necessary to Comply with OSHA standards with this device.
- An innovative water catcher for flushing the emergency chemical shower
Because many lab drains are not ideally located or equipped to contain all the water released during an emergency flushing, this system ensures the water flows at a steady pace that all drains can easily accommodate.
An excellent complement to the shower tester (Wards® 64990-03), this shower flush system includes an elbow valve, flex hose, drain cover and hole cutter for the garbage can (not included). Developed by Joel Palmer, system also includes a lab safety checklist to help you complete the proper testing and maintenance of your safety systems.
Customers Who Bought This Also Bought