Sharpening Stone
Catalog # 470150-112
Supplier: REXEL USA
Specifications
- Description:Sharpening Stone
- Cat. no.:470150-112
- Supplier No.:NOR61463687565
Specifications
About this item
Prolong the life of your important dissecting instruments by sharpening them often on this ultrafine grit sharpening stone (Arkansas novaculite).
Size: 3"L x 1"W
Customers Who Bought This Also Bought